siRNA Details
Gene Symbol: ZNF286A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130842.1 | NP_001124314.1 | 3, 4 | 530 | ||
NM_001288642.1 | NP_001275571.1 | 3, 4 | 457 | |||
NM_001288643.1 | NP_001275572.1 | 2, 3 | 298 | |||
NM_001288645.1 | NP_001275574.1 | 3, 4 | 617 | |||
NM_001288646.1 | NP_001275575.1 | 3, 4 | 530 | |||
NM_001288648.1 | NP_001275577.1 | 2, 3 | 298 | |||
NM_020652.2 | NP_065703.1 | 3, 4 | 617 | |||
GenBank mRNA | AF086305.1 | - | 1, 2 | 50 | ||
AF217226.1 | - | 2, 3 | 449 | |||
AK092259.1 | - | 3, 4 | 455 | |||
AK315062.1 | - | 3, 4 | 375 | |||
BC015730.1 | - | 3, 4 | 455 | |||
BC025260.1 | - | 3, 4 | 455 | |||
BC128259.1 | - | 2, 3 | 285 | 40116891 | ||
BC128411.1 | - | 2, 3 | 285 | 40116881 | ||
DA096356.1 | - | 2, 3 | 121 | |||
DA748526.1 | - | 3, 4 | 368 | |||
DA796080.1 | - | 3, 4 | 290 | |||
DC344287.1 | - | 2, 3 | 184 | |||
DN997307.1 | - | 2, 3 | 313 |
