siRNA Details
Gene Symbol: MTUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020749.4 | NP_065800.1 | 1 | 456 | ||
GenBank mRNA | AF121259.1 | - | 1 | 96 | ||
AF293357.1 | - | 1 | 369 | |||
AK289750.1 | - | 1 | 455 | |||
AK314692.1 | - | 1 | 390 | |||
BC117564.1 | - | 1 | 134 | |||
BC142971.1 | - | 1 | 389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
