siRNA Details
Gene Symbol: TCL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028288.1 | 7 | 1680 | |||
GenBank mRNA | AB035332.1 | - | 4 | 3459 | ||
AB035333.1 | - | 3 | 3323 | |||
AB035334.1 | - | 5 | 2491 | |||
AB035335.1 | - | 4 | 2036 | |||
AB035337.1 | - | 7 | 1682 | |||
AB035338.1 | - | 7 | 1877 | |||
AB035339.1 | - | 2 | 148 | |||
AF195821.1 | - | 3 | 282 |
