siRNA Details
Gene Symbol: Olr1247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000807.1 | NP_001000807.1 | 1 | 227 |
Gene Symbol: LOC100912186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749663.1 | XP_003749711.1 | 1 | 227 |
