siRNA Details
Gene Symbol: Olr1246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000808.1 | NP_001000808.1 | 1 | 902 |
Gene Symbol: LOC100912152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762661.1 | XP_008760883.1 | 1 | 902 |
