siRNA Details
Gene Symbol: Olr921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001385.1 | NP_001001385.1 | 1 | 846 |
Gene Symbol: LOC103689923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765069.1 | XP_008763291.1 | 1 | 846 |
