siRNA Details
Gene Symbol: Klhl36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017511.1 | NP_001017511.1 | 3 | 656 | ||
XM_006255718.2 | XP_006255780.1 | 3 | 768 | |||
XM_008772641.1 | XP_008770863.1 | 3 | 2018 | |||
GenBank mRNA | BC081914.1 | - | 3 | 656 | 7112173 |
