siRNA Details
Gene Symbol: Creld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024783.1 | NP_001019954.1 | 11 | 1519 | ||
XM_006237027.2 | XP_006237089.1 | 10 | 2006 | |||
XM_006237028.2 | XP_006237090.1 | 10 | 1564 | |||
GenBank mRNA | BC097951.1 | - | 11 | 1519 | 7442421 |
