siRNA Details
Gene Symbol: LYZL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199951.2 | NP_001186880.1 | 5 | 827 | ||
NM_020426.3 | NP_065159.1 | 5 | 941 | |||
GenBank mRNA | AY359018.1 | - | 4 | 745 | ||
AY742214.1 | - | 4 | 717 | |||
BC054481.1 | - | 5 | 787 | 6620264 | ||
GQ472214.1 | - | 5 | 787 |
Gene Symbol: LRRC37A11P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033753.2 | 5 | 3457 | |||
GenBank mRNA | AK125814.1 | - | 5 | 3456 | ||
BC111725.1 | - | 5 | 3183 | 40028479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
