siRNA Details
Gene Symbol: ASPHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020437.4 | NP_065170.2 | 4 | 3125 | ||
GenBank mRNA | AK097157.1 | - | 3 | 2470 | ||
AL161993.1 | - | 3 | 2185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
