siRNA Details
Gene Symbol: ACTR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040135.2 | NP_001035225.1 | 7 | 689 | ||
NM_020445.5 | NP_065178.1 | 7 | 689 | |||
NR_073000.1 | 6 | 633 | ||||
NR_073001.1 | 6 | 564 | ||||
XM_006716065.2 | XP_006716128.1 | 7 | 671 | |||
XM_006716066.1 | XP_006716129.1 | 7 | 671 | |||
XM_006716067.3 | XP_006716130.1 | 7 | 671 | |||
XM_011516425.2 | XP_011514727.1 | 7 | 671 | |||
XM_011516426.2 | XP_011514728.1 | 7 | 671 | |||
XM_011516427.1 | XP_011514729.1 | 7 | 671 | |||
XM_017012460.1 | XP_016867949.1 | 7 | 671 | |||
XM_017012461.1 | XP_016867950.1 | 7 | 671 | |||
XM_017012462.1 | XP_016867951.1 | 7 | 600 | |||
XM_017012463.1 | XP_016867952.1 | 7 | 539 | |||
XM_017012464.1 | XP_016867953.1 | 6 | 630 | |||
XM_017012465.1 | XP_016867954.1 | 6 | 493 | |||
GenBank mRNA | AB209174.1 | - | 3 | 4016 | ||
AF023453.1 | - | 7 | 591 | |||
AF086920.1 | - | 6 | 555 | |||
AK001198.1 | - | 3 | 157 | |||
AK294292.1 | - | 6 | 587 | |||
AK315534.1 | - | 7 | 671 | |||
AK316078.1 | - | 6 | 484 | |||
BC008682.1 | - | 7 | 644 | 3854008 | ||
BC015207.1 | - | 7 | 601 | 3916310 | ||
DN601600.1 | - | 7 | 689 |
Gene Symbol: ACTR3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164458.1 | NP_001157930.1 | 4 | 420 | ||
NM_001164459.1 | NP_001157931.1 | 4 | 358 | |||
XM_005250043.4 | XP_005250100.3 | 4 | 420 | |||
XM_011516502.2 | XP_011514804.1 | 4 | 420 | |||
XM_011516506.2 | XP_011514808.1 | 4 | 420 | |||
XM_011516507.2 | XP_011514809.1 | 4 | 420 | |||
XM_011516508.2 | XP_011514810.1 | 4 | 420 | |||
XM_011516511.2 | XP_011514813.1 | 4 | 420 | |||
XM_017012548.1 | XP_016868037.1 | 4 | 420 | |||
XM_017012549.1 | XP_016868038.1 | 4 | 420 | |||
XM_017012550.1 | XP_016868039.1 | 4 | 420 | |||
XM_017012551.1 | XP_016868040.1 | 4 | 420 | |||
XM_017012552.1 | XP_016868041.1 | 4 | 2267 | |||
XM_017012553.1 | XP_016868042.1 | 3, 4 | 2270 | |||
GenBank mRNA | AB039791.1 | - | 4 | 358 | ||
BX640643.1 | - | 3 | 162 | |||
DA868104.1 | - | 4 | 420 | |||
DN992136.1 | - | 4 | 484 | |||
DR004860.1 | - | 4 | 261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
