siRNA Details
Gene Symbol: BC080695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007579.3 | NP_001007580.1 | 537 | |||
GenBank mRNA | BC080695.1 | - | 538 | 30458858 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC080695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007579.3 | NP_001007580.1 | 537 | |||
GenBank mRNA | BC080695.1 | - | 538 | 30458858 |