siRNA Details
Gene Symbol: Olfr456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011528.2 | NP_001011528.2 | 1 | 36 | ||
GenBank mRNA | BC139108.1 | - | 1 | 36 | 8862130 | |
BC139109.1 | - | 1 | 36 | 8862131 |
