siRNA Details
Gene Symbol: Gm527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025605.1 | NP_001020776.1 | 5 | 1076 | ||
GenBank mRNA | BC099503.1 | - | 5 | 1076 | 30940052 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025605.1 | NP_001020776.1 | 5 | 1076 | ||
GenBank mRNA | BC099503.1 | - | 5 | 1076 | 30940052 |