siRNA Details
Gene Symbol: Zswim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029912.2 | NP_001025083.1 | 14 | 5350 | ||
GenBank mRNA | AK076742.1 | - | 1 | 834 | ||
AK122519.1 | - | 14 | 5354 | |||
BC089375.1 | - | 3 | 2764 |

More Information
Other Species Targeted

Gene Ontology Categories:
