siRNA Details
Gene Symbol: Nlrc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033367.3 | NP_001028539.1 | 3, 4 | 470 | ||
GenBank mRNA | AK143864.1 | - | 3, 4 | 471 | ||
AK171624.1 | - | 3, 4 | 465 | |||
BC144861.1 | - | 1, 2 | 241 |
