siRNA Details
Gene Symbol: Slfn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF099975.1 | - | 1 | 770 | ||
AF099976.1 | - | 2 | 1862 | |||
AK137559.1 | - | 8 | 3508 | |||
AK137689.1 | - | 7 | 3429 | |||
BC044865.1 | - | 8 | 3516 | 4459564 | ||
BC046511.1 | - | 3 | 1395 | |||
BC150770.1 | - | 5 | 3123 | 9087681 |
