siRNA Details
Gene Symbol: Xlr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081643.1 | NP_001075112.1 | 2 | 117 | ||
GenBank mRNA | AK146398.1 | - | 2 | 86 | ||
BC038862.1 | - | 2 | 80 | |||
BC057003.1 | - | 2 | 89 | |||
BC064787.1 | - | 2 | 74 | 30046437 | ||
CJ063846.1 | - | 2 | 118 |
Gene Symbol: Xlr3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011727.2 | NP_035857.2 | 2 | 86 | ||
GenBank mRNA | L22977.1 | - | 71 | |||
U02600.1 | - | 2 | 86 |
