siRNA Details
Gene Symbol: INTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020748.2 | NP_065799.1 | 25 | 4759 | ||
NR_026641.1 | 25 | 5040 | ||||
XM_006721987.3 | XP_006722050.1 | 25 | 4766 | |||
XM_011525058.2 | XP_011523360.1 | 25 | 5046 | |||
GenBank mRNA | AB033113.1 | - | 25 | 4987 | ||
AL136800.1 | - | 25 | 4759 |
