siRNA Details
Gene Symbol: 1700034E13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030097.1 | NP_084373.1 | 4 | 343 | ||
GenBank mRNA | AK018840.1 | - | 3 | 303 | ||
BC048563.1 | - | 4 | 343 | 6704020 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700034E13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030097.1 | NP_084373.1 | 4 | 343 | ||
GenBank mRNA | AK018840.1 | - | 3 | 303 | ||
BC048563.1 | - | 4 | 343 | 6704020 |