siRNA Details
Gene Symbol: Vmn1r189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145844.1 | NP_665843.1 | 1 | 73 | ||
GenBank mRNA | BC150715.1 | - | 1 | 172 | 9087626 |
Gene Symbol: Vmn1r221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167542.1 | NP_001161014.1 | 1 | 73 |
