siRNA Details
Gene Symbol: KIAA1468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020854.3 | NP_065905.2 | 8 | 1513 | ||
XM_005266730.2 | XP_005266787.1 | 8 | 1560 | |||
XM_011526110.1 | XP_011524412.1 | 8 | 1560 | |||
XM_011526111.1 | XP_011524413.1 | 8 | 1560 | |||
XM_011526112.1 | XP_011524414.1 | 8 | 1560 | |||
XM_011526114.1 | XP_011524416.1 | 8 | 1560 | |||
XM_017025882.1 | XP_016881371.1 | 8 | 1560 | |||
XM_017025883.1 | XP_016881372.1 | 8 | 1560 | |||
GenBank mRNA | AB040901.1 | - | 4 | 628 | ||
AK091160.1 | - | 8 | 1513 | |||
AK315400.1 | - | 4 | 457 | |||
BC011992.1 | - | 5 | 614 | 4339646 | ||
BX648481.1 | - | 8 | 1497 |
