siRNA Details
Gene Symbol: Zcchc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153160.4 | NP_694800.1 | 7 | 578 | ||
GenBank mRNA | AJ272345.1 | - | 7 | 534 | ||
AK012953.1 | - | 7 | 592 | |||
AK152289.1 | - | 7 | 579 | |||
AK161588.1 | - | 7 | 573 | |||
AK168912.1 | - | 579 | ||||
AY253297.1 | - | 7 | 587 | |||
BC018382.1 | - | 6 | 516 | 4215774 | ||
BC049231.1 | - | 7 | 554 | 4161178 |
