siRNA Details
Gene Symbol: Zfp934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162911.1 | NP_001156383.1 | 4 | 539 | ||
NM_175252.4 | NP_780461.2 | 5 | 743 | |||
GenBank mRNA | AK020133.1 | - | 3 | 508 | ||
AK030776.1 | - | 5 | 746 | |||
BC021319.1 | - | 458 | ||||
BC115861.1 | - | 3 | 390 | |||
BC139791.1 | - | 4 | 451 |
Gene Symbol: 2810408B13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013043.1 | - | 941 |
Gene Symbol: Zfp808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039239.2 | NP_001034328.2 | 4 | 565 | ||
GenBank mRNA | BC061203.1 | - | 4 | 565 | 30248220 |
