siRNA Details
Gene Symbol: Hnrnpa2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016806.3 | NP_058086.2 | 9 | 1111 | ||
NM_182650.4 | NP_872591.1 | 8 | 991 | |||
GenBank mRNA | AF073993.1 | - | 9 | 989 | ||
AF406651.2 | - | 8 | 900 | |||
AK089012.1 | - | 8 | 942 | |||
AK137214.1 | - | 8 | 992 | |||
BC059107.1 | - | 6 | 527 | |||
BC141253.1 | - | 8 | 825 | 9055865 | ||
BC145588.1 | - | 9 | 945 | 9054154 | ||
CN665325.1 | - | 3 | 189 |
