siRNA Details
Gene Symbol: DHRS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282987.1 | NP_001269916.1 | 3 | 387 | ||
NM_001282989.1 | NP_001269918.1 | 3 | 387 | |||
NM_021004.3 | NP_066284.2 | 3 | 387 | |||
XM_006720005.3 | XP_006720068.1 | 3 | 387 | |||
XM_011536366.2 | XP_011534668.1 | 3 | 387 | |||
GenBank mRNA | AB045131.1 | - | 3 | 412 | ||
AF044127.1 | - | 3 | 358 | |||
AF064256.1 | - | 3 | 361 | |||
AK001870.1 | - | 3 | 353 | |||
AK301201.1 | - | 3 | 333 | |||
AK308436.1 | - | 3 | 352 | |||
AK314448.1 | - | 3 | 352 | |||
AY071856.1 | - | 3 | 338 | |||
AY358638.1 | - | 3 | 368 | |||
AY616182.1 | - | 3 | 387 | |||
BC003019.1 | - | 3 | 335 | 2822884 | ||
BP366505.1 | - | 3 | 391 | |||
BU529016.1 | - | 3 | 366 | |||
DQ344810.1 | - | 3 | 366 |
Gene Symbol: DHRS4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193636.1 | NP_001180565.1 | 3 | 664 | ||
NM_001193637.1 | NP_001180566.1 | 3 | 664 | |||
NM_198083.3 | NP_932349.2 | 3 | 463 | |||
GenBank mRNA | AY616183.1 | - | 599 | |||
BC000663.2 | - | 3 | 371 | 3349226 | ||
DN237881.1 | - | 3 | 351 | |||
DN237882.1 | - | 4 | 405 | |||
GU564235.1 | - | 3 | 431 | |||
KC237375.1 | - | 4 | 717 | |||
KU141377.1 | - | 4 | 691 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
