siRNA Details
Gene Symbol: 1700016K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198637.2 | NP_941039.1 | 2 | 544 | ||
GenBank mRNA | AK132806.1 | - | 2 | 469 | ||
BC049758.1 | - | 2 | 483 | 6773542 | ||
BQ931096.1 | - | 2 | 559 |
