siRNA Details
Gene Symbol: Olfr1371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207253.1 | NP_997136.1 | 1 | 754 | ||
GenBank mRNA | BC106788.1 | - | 1 | 754 | 40041434 | |
BC106789.1 | - | 1 | 753 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207253.1 | NP_997136.1 | 1 | 754 | ||
GenBank mRNA | BC106788.1 | - | 1 | 754 | 40041434 | |
BC106789.1 | - | 1 | 753 |