siRNA Details
Gene Symbol: Vmn1r59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207543.1 | NP_997426.1 | 1 | 523 | ||
GenBank mRNA | BC114347.2 | - | 1 | 517 | ||
BC148233.1 | - | 602 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207543.1 | NP_997426.1 | 1 | 523 | ||
GenBank mRNA | BC114347.2 | - | 1 | 517 | ||
BC148233.1 | - | 602 |