siRNA Details
Gene Symbol: ZNFX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021035.2 | NP_066363.1 | 14 | 6296 | ||
XM_006723836.1 | XP_006723899.1 | 14 | 6126 | |||
XM_011528924.1 | XP_011527226.1 | 13 | 6171 | |||
XM_017027971.1 | XP_016883460.1 | 12 | 5222 | |||
GenBank mRNA | AB037825.2 | - | 14 | 6114 | ||
AK000573.1 | - | 1 | 965 | |||
AK023836.1 | - | 10 | 4048 | |||
BC062347.1 | - | 1 | 1635 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
