siRNA Details
Gene Symbol: NDUFV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021075.3 | NP_066553.3 | 3 | 350 | ||
XM_011529586.2 | XP_011527888.1 | 3 | 347 | |||
GenBank mRNA | AK056041.1 | - | 5 | 807 | ||
AK308228.1 | - | 3 | 307 | |||
BC021217.2 | - | 3 | 337 | 4302265 | ||
BF311986.1 | - | 3 | 295 | |||
BI829092.1 | - | 3 | 353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
