siRNA Details
Gene Symbol: WDFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020830.4 | NP_065881.1 | 6, 7 | 700 | ||
GenBank mRNA | AB037856.1 | - | 6, 7 | 613 | ||
AF411977.1 | - | 6, 7 | 597 | |||
AJ310568.1 | - | 6, 7 | 687 | |||
AK022888.1 | - | 6, 7 | 626 | |||
AK023415.1 | - | 5, 6 | 494 | |||
BC040525.1 | - | 6, 7 | 612 | 5285567 | ||
BC065934.1 | - | 6, 7 | 644 | 6164567 |
