siRNA Details
Gene Symbol: PRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020956.2 | NP_066007.1 | 6 | 1064 | ||
GenBank mRNA | AF321192.1 | - | 6 | 1064 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
