siRNA Details
Gene Symbol: SPRR2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014291.3 | NP_001014313.1 | 2 | 635 | ||
XM_017002177.1 | XP_016857666.1 | 4 | 1012 | |||
XM_017002178.1 | XP_016857667.1 | 2 | 558 |
