siRNA Details
Gene Symbol: DHRS4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193635.1 | NP_001180564.1 | 5 | 546 | ||
NM_001193636.1 | NP_001180565.1 | 4 | 789 | |||
NM_198083.3 | NP_932349.2 | 4 | 588 | |||
GenBank mRNA | AK021587.1 | - | 1 | 179 | ||
BC000663.2 | - | 4 | 496 | 3349226 | ||
DQ088987.1 | - | 2 | 87 | |||
DQ088988.1 | - | 2 | 87 | |||
GU564234.1 | - | 5 | 454 | |||
GU564235.1 | - | 4 | 556 |
Gene Symbol: DHRS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282988.1 | NP_001269917.1 | 3 | 410 | ||
NM_001282990.1 | NP_001269919.1 | 3 | 410 | |||
NM_021004.3 | NP_066284.2 | 4 | 512 | |||
XM_006720005.3 | XP_006720068.1 | 4 | 512 | |||
GenBank mRNA | AB045131.1 | - | 4 | 537 | ||
AF044127.1 | - | 4 | 483 | |||
AF064256.1 | - | 4 | 486 | |||
AK001870.1 | - | 4 | 478 | |||
AK314448.1 | - | 4 | 477 | |||
AY358638.1 | - | 4 | 493 | |||
BC003019.1 | - | 4 | 460 | 2822884 | ||
BP366505.1 | - | 4 | 516 | |||
DQ325464.1 | - | 3 | 377 | |||
DQ338571.1 | - | 2 | 239 | |||
DQ344810.1 | - | 4 | 491 |
Gene Symbol: DHRS4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102693.1 | 3 | 528 | |||
XM_006720246.3 | XP_006720309.1 | 3 | 373 | |||
GenBank mRNA | BC171918.1 | - | 3 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
