siRNA Details
Gene Symbol: PSG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021016.3 | NP_066296.2 | 7 | 1561 | ||
XM_011527126.1 | XP_011525428.1 | 6 | 1282 | |||
XM_011527127.1 | XP_011525429.1 | 5 | 1027 | |||
GenBank mRNA | BC005924.1 | - | 7 | 1526 | 4043628 | |
M23575.1 | - | 7 | 1660 | |||
M27907.1 | - | 4 | 644 | |||
M30628.1 | - | 9 | 1589 | |||
M34420.1 | - | 7 | 1510 | |||
M37399.1 | - | 7 | 1515 | |||
M93061.1 | - | 7 | 1487 | |||
S59493.1 | - | 7 | 1487 | |||
S59494.1 | - | 9 | 1595 |
Gene Symbol: PSG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113410.1 | NP_001106881.1 | 5 | 824 | ||
NM_002785.2 | NP_002776.3 | 6 | 1190 | |||
NM_203287.1 | NP_976032.2 | 5 | 892 | |||
GenBank mRNA | BC020711.1 | - | 5 | 882 | 4732286 | |
BE767872.1 | - | 3 | 293 | |||
BG571293.1 | - | 4 | 532 | |||
M69245.1 | - | 6 | 1152 | |||
U25988.1 | - | 5 | 820 |
Gene Symbol: PSG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130014.1 | NP_001123486.1 | 6 | 1214 | ||
NM_002781.3 | NP_002772.3 | 6 | 1282 | |||
XM_017027003.1 | XP_016882492.1 | 6 | 1276 | |||
GenBank mRNA | BC012607.1 | - | 6 | 1249 | 4043912 | |
BG621332.1 | - | 4 | 704 | |||
M25384.1 | - | 7 | 1182 | |||
M73713.1 | - | 6 | 1168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
