siRNA Details
Gene Symbol: ZNF142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105537.2 | NP_001099007.1 | 6 | 1493 | ||
NR_073599.1 | 7 | 1802 | ||||
NR_073600.1 | 7 | 1635 | ||||
XM_006712730.3 | XP_006712793.1 | 7 | 2168 | |||
XM_011511784.2 | XP_011510086.1 | 7 | 2263 | |||
XM_011511785.2 | XP_011510087.1 | 7 | 2188 | |||
XM_011511786.2 | XP_011510088.1 | 7 | 2093 | |||
XM_011511787.2 | XP_011510089.1 | 6 | 2088 | |||
XM_011511788.2 | XP_011510090.1 | 6 | 1993 | |||
XM_011511789.2 | XP_011510091.1 | 6 | 1661 | |||
XM_017004871.1 | XP_016860360.1 | 6 | 2244 | |||
XM_017004872.1 | XP_016860361.1 | 5 | 1644 | |||
GenBank mRNA | BC033631.1 | - | 6 | 2216 | ||
BC053849.2 | - | 7 | 1821 | |||
D87073.1 | - | 6 | 1491 |
