siRNA Details
Gene Symbol: FAM135A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162529.1 | NP_001156001.1 | 13 | 1555 | ||
XM_005248743.1 | XP_005248800.1 | 16 | 1936 | |||
XM_005248744.4 | XP_005248801.1 | 15 | 1903 | |||
XM_005248745.2 | XP_005248802.1 | 15 | 1858 | |||
XM_005248749.1 | XP_005248806.1 | 16 | 1936 | |||
XM_011535993.2 | XP_011534295.1 | 15 | 1656 | |||
XM_011535994.2 | XP_011534296.1 | 16 | 2076 | |||
XM_011535995.2 | XP_011534297.1 | 16 | 1877 | |||
XM_011535996.2 | XP_011534298.1 | 16 | 2034 | |||
XM_011535997.2 | XP_011534299.1 | 14 | 1778 | |||
XM_017011124.1 | XP_016866613.1 | 15 | 1909 | |||
XM_017011125.1 | XP_016866614.1 | 14 | 1681 | |||
XM_017011126.1 | XP_016866615.1 | 15 | 1995 | |||
XM_017011127.1 | XP_016866616.1 | 14 | 1777 | |||
XM_017011128.1 | XP_016866617.1 | 13 | 1701 | |||
XM_017011135.1 | XP_016866624.1 | 14 | 1757 | |||
XM_017011136.1 | XP_016866625.1 | 5 | 484 | |||
GenBank mRNA | AB037832.1 | - | 13 | 1523 | ||
AK000183.1 | - | 12 | 1689 | |||
AK225230.1 | - | 12 | 1689 | |||
AL136820.1 | - | 2 | 180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
