siRNA Details
Gene Symbol: ZNF444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018337.3 | NP_060807.2 | 4, 5 | 791 | ||
XM_005259034.2 | XP_005259091.1 | 5, 6 | 896 | |||
XM_005259035.1 | XP_005259092.1 | 4, 5 | 828 | |||
XM_005259036.1 | XP_005259093.1 | 4, 5 | 695 | |||
XM_011527068.1 | XP_011525370.1 | 4, 5 | 854 | |||
XM_011527069.1 | XP_011525371.1 | 5, 6 | 746 | |||
XM_011527070.1 | XP_011525372.1 | 5, 6 | 2440 | |||
XM_011527071.1 | XP_011525373.1 | 5, 6 | 867 | |||
XM_017026932.1 | XP_016882421.1 | 5, 6 | 729 | |||
XM_017026933.1 | XP_016882422.1 | 2, 3 | 1040 | |||
GenBank mRNA | AB052955.1 | - | 4, 5 | 634 | ||
AK074063.1 | - | 1, 2 | 4346 | |||
AK097319.1 | - | 1, 2 | 2019 | |||
AK223408.1 | - | 4, 5 | 661 | |||
AL834130.1 | - | 4, 5 | 771 |
