siRNA Details
Gene Symbol: DHRS4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277864.1 | NP_001264793.1 | 4 | 876 | ||
NR_102687.1 | 3 | 611 | ||||
NR_102688.1 | 4 | 789 | ||||
NR_102689.1 | 4 | 553 | ||||
NR_102691.1 | 3 | 375 | ||||
NR_102692.1 | 4 | 636 | ||||
NR_102693.1 | 5 | 732 | ||||
NR_102694.1 | 3 | 501 | ||||
XM_006720246.3 | XP_006720309.1 | 6 | 604 | |||
GenBank mRNA | BC171914.1 | - | 4 | 595 | ||
BC171918.1 | - | 5 | 691 | |||
GQ871921.1 | - | 3 | 388 | |||
GQ871922.1 | - | 3 | 348 | |||
GQ871923.1 | - | 4 | 552 | |||
GQ871924.1 | - | 4 | 825 | |||
GQ871925.1 | - | 4 | 567 | |||
GQ871926.1 | - | 3 | 412 | |||
KU058702.1 | - | 4 | 813 | |||
KU058703.1 | - | 4 | 752 | |||
KU058704.1 | - | 4 | 730 | |||
KU058705.1 | - | 4 | 680 | |||
KU058706.1 | - | 3 | 678 | |||
KU058707.1 | - | 3 | 552 | |||
KU058708.1 | - | 4 | 534 | |||
KU058709.1 | - | 2 | 417 |
Gene Symbol: DHRS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282987.1 | NP_001269916.1 | 5 | 537 | ||
NM_001282988.1 | NP_001269917.1 | 6 | 641 | |||
NM_001282989.1 | NP_001269918.1 | 4 | 485 | |||
NM_001282990.1 | NP_001269919.1 | 5 | 506 | |||
NM_001282991.1 | NP_001269920.1 | 3 | 383 | |||
NM_021004.3 | NP_066284.2 | 7 | 743 | |||
XM_006720005.3 | XP_006720068.1 | 6 | 608 | |||
GenBank mRNA | AB045131.1 | - | 7 | 768 | ||
AF044127.1 | - | 7 | 714 | |||
AF064256.1 | - | 7 | 717 | |||
AK001870.1 | - | 7 | 709 | |||
AK301201.1 | - | 4 | 431 | |||
AK308436.1 | - | 4 | 450 | |||
AK314448.1 | - | 7 | 708 | |||
AY071856.1 | - | 4 | 436 | |||
AY358638.1 | - | 7 | 724 | |||
AY616182.1 | - | 5 | 537 | |||
BC003019.1 | - | 7 | 691 | 2822884 | ||
CD674505.1 | - | 3 | 355 | |||
DQ325464.1 | - | 6 | 608 | |||
DQ338571.1 | - | 4 | 335 | |||
DQ344810.1 | - | 7 | 722 |
Gene Symbol: DHRS4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193635.1 | NP_001180564.1 | 8 | 776 | ||
NM_001193636.1 | NP_001180565.1 | 7 | 1019 | |||
NM_001193637.1 | NP_001180566.1 | 5 | 896 | |||
NM_198083.3 | NP_932349.2 | 7 | 818 | |||
GenBank mRNA | AK301373.1 | - | 4 | 523 | ||
AY616183.1 | - | 3 | 697 | |||
AY943857.1 | - | 3 | 595 | |||
BC000663.2 | - | 7 | 726 | 3349226 | ||
DN237882.1 | - | 5 | 503 | |||
DQ088987.1 | - | 5 | 317 | |||
GU564234.1 | - | 8 | 685 | |||
GU564235.1 | - | 7 | 787 | |||
KC237375.1 | - | 5 | 815 | |||
KU141377.1 | - | 5 | 789 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
