siRNA Details
Gene Symbol: PPP2R3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013239.4 | NP_037371.2 | 11 | 1681 | ||
XM_011545630.2 | XP_011543932.1 | 11 | 1681 | |||
GenBank mRNA | AB209137.1 | - | 7 | 1287 | ||
AF135016.1 | - | 10 | 1045 | |||
AF155098.1 | - | 5 | 456 | |||
AL157501.1 | - | 3 | 392 | |||
BC009032.1 | - | 3 | 638 | |||
BC011180.1 | - | 5 | 458 | |||
BC063429.1 | - | 11 | 1436 | 5239974 | ||
BF530463.1 | - | 3 | 645 | |||
BM715684.1 | - | 5 | 421 |
