siRNA Details
Gene Symbol: NFS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021100.4 | NP_066923.3 | 5 | 513 | ||
NR_037570.2 | 6 | 635 | ||||
GenBank mRNA | AF097025.2 | - | 5 | 499 | ||
AJ010952.1 | - | 1 | 21 | |||
AK001265.1 | - | 5 | 499 | |||
AK001470.1 | - | 5 | 509 | |||
AK056242.1 | - | 5 | 784 | |||
AK223239.1 | - | 5 | 499 | |||
AK295215.1 | - | 5 | 500 | |||
AK297969.1 | - | 6 | 622 | |||
AK301969.1 | - | 5 | 497 | |||
AK312293.1 | - | 5 | 500 | |||
BC065560.1 | - | 5 | 485 | 6101221 | ||
DA082829.1 | - | 5 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
