siRNA Details
Gene Symbol: SEPT5-GP1BB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037611.1 | 12 | 4521 | |||
NR_037612.1 | 12 | 3025 | ||||
GenBank mRNA | BC024304.1 | - | 5 | 2182 | ||
L20860.1 | - | 1 | 2032 | |||
U59632.1 | - | 12 | 3024 |
Gene Symbol: GP1BB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000407.4 | NP_000398.1 | 2 | 808 | ||
GenBank mRNA | J03259.1 | - | 2 | 803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
