siRNA Details
Gene Symbol: SUPT6H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320755.1 | NP_001307684.1 | 4, 5 | 551 | ||
NM_003170.4 | NP_003161.2 | 3, 4 | 449 | |||
XM_017024978.1 | XP_016880467.1 | 5, 6 | 610 | |||
XM_017024979.1 | XP_016880468.1 | 3, 4 | 687 | |||
XM_017024980.1 | XP_016880469.1 | 2, 3 | 381 | |||
GenBank mRNA | AL833607.1 | - | 4, 5 | 552 | ||
BC136522.1 | - | 3, 4 | 361 | 9020512 | ||
BC136524.1 | - | 3, 4 | 361 | 9020514 | ||
BC150268.1 | - | 3, 4 | 363 | 9007230 | ||
D79984.1 | - | 3, 4 | 363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
