siRNA Details
Gene Symbol: SLC9A3R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130012.2 | NP_001123484.1 | 7 | 1616 | ||
NM_001252073.1 | NP_001239002.1 | 6 | 1233 | |||
NM_001252075.1 | NP_001239004.1 | 6 | 1227 | |||
NM_001252076.1 | NP_001239005.1 | 6 | 1200 | |||
NM_004785.5 | NP_004776.3 | 7 | 1583 | |||
XM_005255702.4 | XP_005255759.1 | 6 | 2296 | |||
XM_006720981.1 | XP_006721044.1 | 6 | 3093 | |||
XM_017023894.1 | XP_016879383.1 | 6 | 2186 | |||
XM_017023895.1 | XP_016879384.1 | 7 | 1471 | |||
GenBank mRNA | AF004900.1 | - | 7 | 1493 | ||
BC014513.1 | - | 7 | 1590 | |||
BC069014.1 | - | 7 | 1573 | |||
BC106001.1 | - | 7 | 1581 | 5296143 | ||
U82108.1 | - | 7 | 1535 | |||
Z50150.1 | - | 7 | 1569 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
