siRNA Details
Gene Symbol: MAGEA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005362.3 | NP_005353.1 | 3 | 643 | ||
XM_005274676.3 | XP_005274733.1 | 3 | 717 | |||
XM_006724818.2 | XP_006724881.1 | 3 | 799 | |||
XM_011531160.2 | XP_011529462.1 | 3 | 623 | |||
XM_011531161.2 | XP_011529463.1 | 3 | 1048 | |||
GenBank mRNA | AK223519.1 | - | 1 | 470 | ||
AK292384.1 | - | 3 | 643 | |||
BC000340.2 | - | 3 | 608 | 2822978 | ||
BC005963.1 | - | 3 | 643 | 4064879 | ||
BC011744.2 | - | 2 | 569 | 3345801 | ||
BC016803.1 | - | 3 | 605 | 4073332 | ||
BC017389.1 | - | 3 | 600 | 4047542 | ||
CR541767.1 | - | 1 | 434 | |||
CR541774.1 | - | 1 | 434 |
Gene Symbol: MAGEA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC067731.1 | - | 3 | 622 | 4828429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
