siRNA Details
Gene Symbol: MAGEA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005363.3 | NP_005354.1 | 3 | 515 | ||
NM_175868.2 | NP_787064.1 | 3 | 579 | |||
GenBank mRNA | AK289437.1 | - | 3 | 499 | ||
BC041599.1 | - | 3 | 488 | 5458512 | ||
BC067731.1 | - | 3 | 496 | 4828429 | ||
BE542284.1 | - | 3 | 473 | |||
BM015284.1 | - | 3 | 515 | |||
BU182516.1 | - | 3 | 529 | |||
BX640600.1 | - | 3 | 876 | |||
D32076.1 | - | 1 | 373 | |||
EU161101.1 | - | 1 | 308 | |||
U10339.1 | - | 1 | 308 |
Gene Symbol: MAGEA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR541768.1 | - | 1 | 308 | ||
CR542020.1 | - | 1 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
