siRNA Details
Gene Symbol: DYNC1LI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286157.1 | NP_001273086.1 | 6 | 645 | ||
NM_001323955.1 | NP_001310884.1 | 6 | 965 | |||
NM_006141.2 | NP_006132.1 | 7 | 876 | |||
NR_136658.1 | 7 | 1082 | ||||
XM_017023008.1 | XP_016878497.1 | 7 | 875 | |||
GenBank mRNA | AF035812.1 | - | 7 | 851 | ||
AK095237.1 | - | 1 | 1124 | |||
AK291766.1 | - | 7 | 855 | |||
AK295049.1 | - | 7 | 875 | |||
AK303031.1 | - | 6 | 624 | |||
AK304233.1 | - | 6 | 731 | |||
BC025959.1 | - | 7 | 851 | 4329417 | ||
EB387189.1 | - | 4 | 447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
