siRNA Details
Gene Symbol: DYNC1LI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286157.1 | NP_001273086.1 | 8 | 885 | ||
NM_001323955.1 | NP_001310884.1 | 8 | 1205 | |||
NM_006141.2 | NP_006132.1 | 9 | 1116 | |||
NR_136658.1 | 9 | 1322 | ||||
XM_017023008.1 | XP_016878497.1 | 9 | 1115 | |||
GenBank mRNA | AF035812.1 | - | 9 | 1091 | ||
AK095237.1 | - | 2 | 3479 | |||
AK291766.1 | - | 9 | 1095 | |||
AK303031.1 | - | 8 | 864 | |||
BC025959.1 | - | 9 | 1091 | 4329417 | ||
EB387189.1 | - | 6 | 687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
