siRNA Details
Gene Symbol: CBY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002880.1 | NP_001002880.2 | 6 | 855 | ||
NM_015373.3 | NP_056188.1 | 5 | 715 | |||
GenBank mRNA | AB015347.1 | - | 2 | 402 | ||
AF331041.1 | - | 5 | 651 | |||
AL050345.1 | - | 5 | 694 | |||
AL136686.1 | - | 5 | 697 | |||
BC016139.1 | - | 5 | 693 | 3922707 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
